Paracetamol 98%

Supplier: Apollo Scientific
Warning

OR3962-5KG OR3962-1KG OR3962-250G
APOSOR3962-5KGEA 574 EUR
APOSOR3962-5KG APOSOR3962-1KG APOSOR3962-250G
Paracetamol 98%
Paracetamol
Formula: C₈H₉NO₂
Melting Pt: 168…172 °C
Density: 1,293 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002328
CAS Number: 103-90-2
EINECS: 203-157-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR