2-Bromo-4-chlorobenzyl bromide 95%

Supplier: Apollo Scientific

OR400062-25G OR400062-5G
APOSOR400062-25GEA 464 EUR
APOSOR400062-25G APOSOR400062-5G
2-Bromo-4-chlorobenzyl bromide 95%
2-Bromo-4-chlorobenzyl bromide
Formula: C₇H₅Br₂Cl
MW: 284.376 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12024978
CAS Number: 33924-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR