5-Iodo-2-nitroanisole

Supplier: Apollo Scientific
Warning

OR400064-1G OR400064-5G
APOSOR400064-1GEA 92.8 EUR
APOSOR400064-1G APOSOR400064-5G
5-Iodo-2-nitroanisole
5-Iodo-2-nitroanisole
Formula: C₇H₆INO₃
Melting Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD18397934
CAS Number: 860585-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR