4-Iodo-5-methyl-2-nitroaniline

Supplier: Apollo Scientific
Warning

Synonyms: 2-Amino-5-iodo-4-methylnitrobenzene, 5-Amino-2-iodo-4-nitrotoluene, 4-Iodo-6-nitro-m-toluidine

OR400191-1G OR400191-250MG OR400191-5G
APOSOR400191-1GEA 685 EUR
APOSOR400191-1G APOSOR400191-250MG APOSOR400191-5G
4-Iodo-5-methyl-2-nitroaniline
4-Iodo-5-methyl-2-nitroaniline
Formula: C₇H₇IN₂O₂
Melting Pt: 166…169 °C
Storage Temperature: Ambient
MDL Number: MFCD27949584
CAS Number: 335349-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR