4-Methoxy-2-nitrobenzyl alcohol

Supplier: Apollo Scientific
Warning

OR400470-1G OR400470-250MG
APOSOR400470-1GEA 441 EUR
APOSOR400470-1G APOSOR400470-250MG
4-Methoxy-2-nitrobenzyl alcohol
4-Methoxy-2-nitrobenzyl alcohol
Formula: C₈H₉NO₄
Melting Pt: 79…82 °C
Storage Temperature: Ambient
MDL Number: MFCD05148477
CAS Number: 22996-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR