2-Bromo-3-iodopyridine

Supplier: Apollo Scientific
Danger

OR400475-1G OR400475-25G OR400475-5G
APOSOR400475-1GEA 23.2 EUR
APOSOR400475-1G APOSOR400475-25G APOSOR400475-5G
2-Bromo-3-iodopyridine
2-Bromo-3-iodopyridine
Formula: C₅H₃BrIN
Melting Pt: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD09056782
CAS Number: 265981-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR