2-Bromo-3-iodobenzyl bromide

Supplier: Apollo Scientific
Danger

OR400517-5G OR400517-1G OR400517-250MG
APOSOR400517-5GEA 424 EUR
APOSOR400517-5G APOSOR400517-1G APOSOR400517-250MG
2-Bromo-3-iodobenzyl bromide
2-Bromo-3-iodobenzyl bromide
Formula: C₇H₅Br₂I
Storage Temperature: Ambient
MDL Number: MFCD12025461
CAS Number: 1261649-03-9
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR