Methyl 2-bromo-3-iodobenzoate

Supplier: Apollo Scientific
Warning

OR400525-1G OR400525-5G OR400525-25G
APOSOR400525-1GEA 78.3 EUR
APOSOR400525-1G APOSOR400525-5G APOSOR400525-25G
Methyl 2-bromo-3-iodobenzoate
Methyl 2-bromo-3-iodobenzoate
Formula: C₈H₆BrIO₂
Storage Temperature: Ambient
MDL Number: MFCD11520418
CAS Number: 1261797-04-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR