Methyl 4-Bromo-3-(bromomethyl)benzoate

Supplier: Apollo Scientific
Warning

OR400600-1G OR400600-250MG OR400600-5G
APOSOR400600-1GEA 132 EUR
APOSOR400600-1G APOSOR400600-250MG APOSOR400600-5G
Methyl 4-Bromo-3-(bromomethyl)benzoate
Methyl 4-Bromo-3-(bromomethyl)benzoate
Formula: C₉H₈Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11858300
CAS Number: 142031-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR