Ethyl-4-chloronicotinate

Supplier: Apollo Scientific
Warning

OR40065-1G OR40065-250MG OR40065-5G
APOSOR40065-1GEA 567 EUR
APOSOR40065-1G APOSOR40065-250MG APOSOR40065-5G
Ethyl-4-chloronicotinate
Ethyl-4-chloronicotinate
Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Boiling Pt: 85 °C/1mmHg
Storage Temperature: Ambient
MDL Number: MFCD05664028
CAS Number: 37831-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR