4-Bromo-3-methylbenzyl alcohol, Technical Grade

Supplier: Apollo Scientific
Warning

OR400665-5G
APOSOR400665-5GEA 137 EUR
APOSOR400665-5G
4-Bromo-3-methylbenzyl alcohol, Technical Grade
4-Bromo-3-methylbenzyl alcohol
Formula: C₈H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD09264005
CAS Number: 149104-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR