6-Bromo-3-iodo-2-nitroanisole

Supplier: Apollo Scientific
Warning

OR400889-1G OR400889-250MG OR400889-5G
APOSOR400889-1GEA 1220 EUR
APOSOR400889-1G APOSOR400889-250MG APOSOR400889-5G
6-Bromo-3-iodo-2-nitroanisole
6-Bromo-3-iodo-2-nitroanisole
Formula: C₇H₅BrINO₃
Storage Temperature: Ambient
MDL Number: MFCD30730277
CAS Number: 2149590-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR