2-Chloro-3-nitrobenzaldehyde 95%

Supplier: Apollo Scientific
Warning

OR401003-1G OR401003-5G
APOSOR401003-1GEA 125 EUR
APOSOR401003-1G APOSOR401003-5G
2-Chloro-3-nitrobenzaldehyde 95%
2-Chloro-3-nitrobenzaldehyde
Formula: C₇H₄ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD08236804
CAS Number: 58755-57-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR