2-Iodo-3-nitrobenzyl alcohol 95%

Supplier: Apollo Scientific
Warning

OR401053-1G OR401053-250MG OR401053-5G
APOSOR401053-1GEA 522 EUR
APOSOR401053-1G APOSOR401053-250MG APOSOR401053-5G
2-Iodo-3-nitrobenzyl alcohol 95%
2-Iodo-3-nitrobenzyl alcohol
Formula: C₇H₆INO₃
Storage Temperature: Ambient
MDL Number: MFCD18397984
CAS Number: 158616-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR