5-Methyl-1H-indazole-6-boronic acid

Supplier: Apollo Scientific
Warning

OR40156-1G OR40156-250MG
APOSOR40156-1GEA 78.3 EUR
APOSOR40156-1G APOSOR40156-250MG
5-Methyl-1H-indazole-6-boronic acid
5-Methyl-1H-indazole-6-boronic acid
Formula: C₈H₉BN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD12028648
CAS Number: 1310383-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR