1,5-Dimethyl-1H-indazole-6-boronic acid

Supplier: Apollo Scientific
Warning

OR40167-1G OR40167-250MG
APOSOR40167-1GEA 136 EUR
APOSOR40167-1G APOSOR40167-250MG
1,5-Dimethyl-1H-indazole-6-boronic acid
1,5-Dimethyl-1H-indazole-6-boronic acid
Formula: C₉H₁₁BN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD12028659
CAS Number: 1310383-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR