8-Amino-5-bromoisoquinoline

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromoisoquinolin-8-amine

OR40222-1G
APOSOR40222-1GEA 23.2 EUR
APOSOR40222-1G
8-Amino-5-bromoisoquinoline
8-Amino-5-bromoisoquinoline
Formula: C₉H₇BrN₂
MW: 223.0693 g/mol
Melting Pt: 187…190 °C
Storage Temperature: Ambient
MDL Number: MFCD09261047
CAS Number: 90721-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR