3-Amino-5-chloropyridine 98%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Chloropyridin-3-amine

OR40335-1G OR40335-25G OR40335-5G
APOSOR40335-1GEA 81.2 EUR
APOSOR40335-1G APOSOR40335-25G APOSOR40335-5G
3-Amino-5-chloropyridine 98%
3-Amino-5-chloropyridine
Formula: C₅H₅ClN₂
Melting Pt: 71…75 °C
Storage Temperature: Ambient
MDL Number: MFCD03701386
CAS Number: 22353-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR