5-Methylpyridin-3-ol

Supplier: Apollo Scientific
Warning

Synonyms: 3-Hydroxy-5-methylpyridine, 3-Hydroxy-5-picoline

OR40438-5G
APOSOR40438-5GEA 151 EUR
APOSOR40438-5G
5-Methylpyridin-3-ol
5-Methylpyridin-3-ol
Formula: C₆H₇NO
MW: 109.13 g/mol
Melting Pt: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00661297
CAS Number: 42732-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR