Methyl 3-acetylbenzoate

Supplier: Apollo Scientific
Warning

Synonyms: 1-[3-(Methoxycarbonyl)phenyl]ethan-1-one, 3'-(Methoxycarbonyl)acetophenone

OR40606-1G OR40606-5G OR40606-10G
APOSOR40606-1GEA 21.9 EUR
APOSOR40606-1G APOSOR40606-5G APOSOR40606-10G
Methyl 3-acetylbenzoate
Methyl 3-acetylbenzoate
Formula: C₁₀H₁₀O₃
Melting Pt: 43…46 °C
Storage Temperature: Ambient
MDL Number: MFCD09475799
CAS Number: 21860-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR