4-(Aminomethyl)phenylacetic acid ethyl ester hydrochloride

Supplier: Apollo Scientific
Warning

OR40626-250MG
APOSOR40626-250MGEA 127 EUR
APOSOR40626-250MG
4-(Aminomethyl)phenylacetic acid ethyl ester hydrochloride
4-(Aminomethyl)phenylacetic acid ethyl ester hydrochloride
Formula: C₁₁H₁₆ClNO₂
Melting Pt: 181…186 °C
Storage Temperature: Ambient
MDL Number: MFCD09953123
CAS Number: 17841-69-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR