1-Boc-4-iodopiperidine 97%

Supplier: Apollo Scientific
Warning

OR41125-25G OR41125-5G OR41125-100G
APOSOR41125-25GEA 55.1 EUR
APOSOR41125-25G APOSOR41125-5G APOSOR41125-100G
1-Boc-4-iodopiperidine 97%
1-Boc-4-iodopiperidine
Formula: C₁₀H₁₈INO₂
Melting Pt: 47…49 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115041
CAS Number: 301673-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR