5-Formyl-2-methoxyphenylboronic acid

Supplier: Apollo Scientific
Warning

OR4117-250MG
APOSOR4117-250MGEA 14.7 EUR
APOSOR4117-250MG
5-Formyl-2-methoxyphenylboronic acid
5-Formyl-2-methoxyphenylboronic acid
Formula: C₈H₉BO₄
Melting Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD01319044
CAS Number: 127972-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR