4-Amino-7-bromo-8-methylquinoline

Supplier: Apollo Scientific

Synonyms: 7-Bromo-8-methylquinolin-4-amine

OR42033-1G
APOSOR42033-1GEA 565 EUR
APOSOR42033-1G
4-Amino-7-bromo-8-methylquinoline
4-Amino-7-bromo-8-methylquinoline
Formula: C₁₀H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD12674989
CAS Number: 1189106-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR