8-Chloroquinoline 98%

Supplier: Apollo Scientific
Warning

OR42189-100G
APOSOR42189-100GEA 165 EUR
APOSOR42189-100G
8-Chloroquinoline 98%
8-Chloroquinoline
Formula: C₉H₆ClN
Boiling Pt: 288 °C
Melting Pt: –20 °C
Density: 1,28 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00047618
CAS Number: 611-33-6
EINECS: 210-265-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR