6-Methoxyquinoline-3-carboxylic acid

Supplier: Apollo Scientific

OR42250-1G OR42250-5G
APOSOR42250-1GEA 42.2 EUR
APOSOR42250-1G APOSOR42250-5G
6-Methoxyquinoline-3-carboxylic acid
6-Methoxyquinoline-3-carboxylic acid
Formula: C₁₁H₉NO₃
Melting Pt: 273 °C
Storage Temperature: Ambient
MDL Number: MFCD08437579
CAS Number: 71082-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR