5-Bromo-2-hydroxypyridine

Supplier: Apollo Scientific
Warning

OR4273-25G OR4273-5G
APOSOR4273-25GEA 24.8 EUR
APOSOR4273-25G APOSOR4273-5G
5-Bromo-2-hydroxypyridine
5-Bromo-2-hydroxypyridine
Formula: C₅H₄BrNO
Melting Pt: 174…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00234042
CAS Number: 13466-38-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR