4-Hydroxy-1H-indazole

Supplier: Apollo Scientific
Warning

Synonyms: 1H-Indazol-4-ol

OR43567-250MG
APOSOR43567-250MGEA 37.7 EUR
APOSOR43567-250MG
4-Hydroxy-1H-indazole
4-Hydroxy-1H-indazole
Formula: C₇H₆N₂O
MW: 134.14 g/mol
Melting Pt: 163…165 °C
Storage Temperature: Ambient
MDL Number: MFCD06858736
CAS Number: 81382-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR