2'-Methoxy-3'-methylacetophenone

Supplier: Apollo Scientific
Warning

OR43670-1G OR43670-250MG OR43670-5G
APOSOR43670-1GEA 174 EUR
APOSOR43670-1G APOSOR43670-250MG APOSOR43670-5G
2'-Methoxy-3'-methylacetophenone
2'-Methoxy-3'-methylacetophenone
Formula: C₁₀H₁₂O₂
Boiling Pt: 240 °C
Density: 1,061 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD11044853
CAS Number: 6342-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR