2,5-Dibromo-3-formylpyridine

Supplier: Apollo Scientific

Synonyms: 2,5-Dibromonicotinaldehyde

OR4434-1G OR4434-250MG OR4434-500MG OR4434-2.5G
APOSOR4434-1GEA 314 EUR
APOSOR4434-1G APOSOR4434-250MG APOSOR4434-500MG APOSOR4434-2.5G
2,5-Dibromo-3-formylpyridine
2,5-Dibromo-3-formylpyridine
Formula: C₆H₃Br₂NO
Melting Pt: 75,5…77 °C
Storage Temperature: Ambient
MDL Number: MFCD07368560
CAS Number: 852181-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR