1-(4-Bromo-3-nitrophenyl)-1-butanone 98%

Supplier: Apollo Scientific
Warning

OR450141-250MG OR450141-1G OR450141-5G
APOSOR450141-250MGEA 241 EUR
APOSOR450141-250MG APOSOR450141-1G APOSOR450141-5G
1-(4-Bromo-3-nitrophenyl)-1-butanone 98%
1-(4-Bromo-3-nitrophenyl)-1-butanone
Formula: C₁₀H₁₀BrNO₃
MDL Number: MFCD34474617
CAS Number: 91715-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR