3-(1-Azetidinyl)-pyrrolidine dihydrochloride

Supplier: Apollo Scientific

OR451027-1G
APOSOR451027-1GEA 1540 EUR
APOSOR451027-1G
3-(1-Azetidinyl)-pyrrolidine dihydrochloride
3-(1-Azetidinyl)-pyrrolidine dihydrochloride
Formula: C₇H₁₄N₂·2ClH
MW: 199.121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28975197
CAS Number: 1018443-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR