(R)-3-Methoxypyrrolidine hydrochloride

Supplier: Apollo Scientific
Warning

OR451217-1G OR451217-5G
APOSOR451217-1GEA 85.7 EUR
APOSOR451217-1G APOSOR451217-5G
(R)-3-Methoxypyrrolidine hydrochloride
(R)-3-Methoxypyrrolidine hydrochloride
Formula: C₅H₁₂ClNO
MW: 137.6079 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11849816
CAS Number: 474707-30-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR