1-Amino-1-indancarboxylic acid methyl ester hydrochloride

Supplier: Apollo Scientific

OR451229-1G
APOSOR451229-1GEA 960 EUR
APOSOR451229-1G
1-Amino-1-indancarboxylic acid methyl ester hydrochloride
1-Amino-1-indancarboxylic acid methyl ester hydrochloride
Formula: C₁₁H₁₃NO₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD08461277
CAS Number: 111140-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR