2,4-Dimethylbenzyl alcohol 97%

Supplier: Apollo Scientific
Warning

OR451325-25G OR451325-5G
APOSOR451325-25GEA 108 EUR
APOSOR451325-25G APOSOR451325-5G
2,4-Dimethylbenzyl alcohol 97%
2,4-Dimethylbenzyl alcohol
Formula: C₉H₁₂O
Boiling Pt: 120 °C
Melting Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00004624
CAS Number: 16308-92-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR