3,5-Dibromobenzaldehyde

Supplier: Apollo Scientific
Danger

OR4526-100G OR4526-25G OR4526-5G
APOSOR4526-100GEA 166 EUR
APOSOR4526-100G APOSOR4526-25G APOSOR4526-5G
3,5-Dibromobenzaldehyde
3,5-Dibromobenzaldehyde
Formula: C₇H₄Br₂O
MW: 263.92 g/mol
Melting Pt: 89.7…91.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00156887
CAS Number: 56990-02-4
UN: 3261
ADR: 8 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR