(S)-1-Boc-3-hydroxypiperidine 98%

Supplier: Apollo Scientific
Warning

OR4581-25G OR4581-5G OR4581-100G
APOSOR4581-25GEA 33.5 EUR
APOSOR4581-25G APOSOR4581-5G APOSOR4581-100G
(S)-1-Boc-3-hydroxypiperidine 98%
(S)-1-Boc-3-hydroxypiperidine
Formula: C₁₀H₁₉NO₃
Melting Pt: 34…40 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD04115307
CAS Number: 143900-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR