4-Chloro-5H-pyrrolo[3,2-d]pyrimidine

Supplier: Apollo Scientific
Warning

OR46059-1G
APOSOR46059-1GEA 21.9 EUR
APOSOR46059-1G
4-Chloro-5H-pyrrolo[3,2-d]pyrimidine
4-Chloro-5H-pyrrolo[3,2-d]pyrimidine
Formula: C₆H₄ClN₃
Storage Temperature: Ambient
MDL Number: MFCD06658411
CAS Number: 84905-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR