6-Bromo-4-methylpyridin-2-amine

Supplier: Apollo Scientific
Warning

Synonyms: 2-Amino-6-bromo-4-methylpyridine

OR46205-1G OR46205-5G OR46205-10G
APOSOR46205-1GEA 21.9 EUR
APOSOR46205-1G APOSOR46205-5G APOSOR46205-10G
6-Bromo-4-methylpyridin-2-amine
6-Bromo-4-methylpyridin-2-amine
Formula: C₆H₇BrN₂
MW: 187.05 g/mol
Melting Pt: 115…116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11036206
CAS Number: 73895-98-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR