(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine 98%

Supplier: Apollo Scientific
Warning

OR4642-5G
APOSOR4642-5GEA 21.9 EUR
APOSOR4642-5G
(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine 98%
(3R)-(+)-3-(tert-Butoxycarbonylamino)pyrrolidine
Formula: C₉H₁₈N₂O₂
Melting Pt: 50…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00143191
CAS Number: 122536-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR