6-Methoxy-2-tetralone

Supplier: Apollo Scientific
Warning

Synonyms: 6-Methoxy-3,4-dihydronaphthalen-2(1H)-one, 3,4-Dihydro-6-methoxynaphthalen-2(1H)-one

OR4643-1G OR4643-5G
APOSOR4643-1GEA 183 EUR
APOSOR4643-1G APOSOR4643-5G
6-Methoxy-2-tetralone
6-Methoxy-2-tetralone
Formula: C₁₁H₁₂O₂
Boiling Pt: 114…116 °C/0,2mmHg
Melting Pt: 30…37 °C
Flash Pt: >113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001729
CAS Number: 2472-22-2
EINECS: 219-592-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR