tert-Butyl (2-bromoethyl)carbamate 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-(BOC-amino)ethyl bromide

OR46552-1G OR46552-25G OR46552-5G
APOSOR46552-1GEA 29.2 EUR
APOSOR46552-1G APOSOR46552-25G APOSOR46552-5G
tert-Butyl (2-bromoethyl)carbamate 97%
tert-Butyl (2-bromoethyl)carbamate
Formula: C₇H₁₄BrNO₂
MW: 224.10 g/mol
Melting Pt: 30…32 °C
Storage Temperature: Freezer
MDL Number: MFCD02683428
CAS Number: 39684-80-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR