[1,2-Bis(diphenylphosphino)ethane]palladium(II) dichloride 95%

Supplier: Apollo Scientific
Warning

OR46570-5G
APOSOR46570-5GEA 215 EUR
APOSOR46570-5G
[1,2-Bis(diphenylphosphino)ethane]palladium(II) dichloride 95%
[1,2-Bis(diphenylphosphino)ethane]palladium(II) dichloride
Formula: C₂₆H₂₄Cl₂P₂Pd
Storage Temperature: Ambient
MDL Number: MFCD00015702
CAS Number: 19978-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR