3-Bromo-4,5-dichloropyridine 97%

Supplier: Apollo Scientific

OR46598-250MG OR46598-1G OR46598-5G
APOSOR46598-250MGEA 63.8 EUR
APOSOR46598-250MG APOSOR46598-1G APOSOR46598-5G
3-Bromo-4,5-dichloropyridine 97%
3-Bromo-4,5-dichloropyridine
Formula: C₅H₂BrCl₂N
Storage Temperature: Ambient
MDL Number: MFCD17014545
CAS Number: 1001056-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR