4-(4-Bromophenyl)but-3-en-2-ol

Supplier: Apollo Scientific

OR46700-1G OR46700-50MG OR46700-250MG OR46700-500MG
APOSOR46700-1GEA 780 EUR
APOSOR46700-1G APOSOR46700-50MG APOSOR46700-250MG APOSOR46700-500MG
4-(4-Bromophenyl)but-3-en-2-ol
4-(4-Bromophenyl)but-3-en-2-ol
Formula: C₁₀H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD08457526
CAS Number: 539825-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR