2-Hydroxybutanenitrile

Supplier: Apollo Scientific

OR46723-1G OR46723-250MG OR46723-500MG OR46723-2.5G OR46723-5G
APOSOR46723-1GEA 328 EUR
APOSOR46723-1G APOSOR46723-250MG APOSOR46723-500MG APOSOR46723-2.5G APOSOR46723-5G
2-Hydroxybutanenitrile
2-Hydroxybutanenitrile
Formula: C₄H₇NO
Storage Temperature: Ambient
MDL Number: MFCD00042729
CAS Number: 4476-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR