2-Acetyl-5-chlorothiophene 99%

Supplier: Apollo Scientific
Warning

OR4686-100G OR4686-25G OR4686-500G
APOSOR4686-100GEA 82.8 EUR
APOSOR4686-100G APOSOR4686-25G APOSOR4686-500G
2-Acetyl-5-chlorothiophene 99%
2-Acetyl-5-chlorothiophene
Formula: C₆H₅ClOS
Boiling Pt: 117…118 °C/17mmHg
Melting Pt: 46…49 °C
Flash Pt: 108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005444
CAS Number: 6310-09-4
EINECS: 228-630-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR