Ethyl 6-(Methylsulphonyl)-2-pyridinecarboxylate

Supplier: Apollo Scientific

OR470053-5G
APOSOR470053-5GEA 1120 EUR
APOSOR470053-5G
Ethyl 6-(Methylsulphonyl)-2-pyridinecarboxylate
Ethyl 6-(Methylsulphonyl)-2-pyridinecarboxylate
Formula: C₉H₁₁NO₄S
Storage Temperature: Ambient
MDL Number: MFCD12828711
CAS Number: 1186663-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR