Ethyl 5,8-Dihydroxyimidazo[1,5-a]pyrazine-1-carboxylate

Supplier: Apollo Scientific

OR470165-5G
APOSOR470165-5GEA 1330 EUR
APOSOR470165-5G
Ethyl 5,8-Dihydroxyimidazo[1,5-a]pyrazine-1-carboxylate
Ethyl 5,8-Dihydroxyimidazo[1,5-a]pyrazine-1-carboxylate
Formula: C₉H₉N₃O₄
MW: 223,185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18071049
CAS Number: 1256633-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR