C-[1-(4-Chlorophenyl)cyclopentyl]methylamine

Supplier: Apollo Scientific

OR470260-5G
APOSOR470260-5GEA 1410 EUR
APOSOR470260-5G
C-[1-(4-Chlorophenyl)cyclopentyl]methylamine
C-[1-(4-Chlorophenyl)cyclopentyl]methylamine
Formula: C₁₂H₁₆ClN
Storage Temperature: Ambient
MDL Number: MFCD07801114
CAS Number: 75180-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR